Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ste2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007718.2 | NP_001007719.1 | 3 | 268 | ||
XM_006250793.2 | XP_006250855.1 | 4 | 559 | |||
XM_008770039.1 | XP_008768261.1 | 5 | 473 | |||
GenBank mRNA | BC088157.1 | - | 3 | 268 | 7323209 | |
S76490.1 | - | 3 | 191 | |||
U50205.1 | - | 2 | 161 |