Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019249.1 | NP_062122.1 | 13 | 2885 | ||
XM_006238677.2 | XP_006238739.1 | 16 | 3928 | |||
XM_006238678.2 | XP_006238740.1 | 15 | 3901 | |||
XM_006238680.2 | XP_006238742.1 | 15 | 3901 | |||
XM_006238681.2 | XP_006238743.1 | 14 | 3595 | |||
GenBank mRNA | L11586.1 | - | 13 | 2885 |