Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1305733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014114.1 | NP_001014136.1 | 642 | |||
GenBank mRNA | BC089943.1 | - | 642 | 7311457 |