Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004661.3 | NP_004652.2 | 10 | 1132 | ||
GenBank mRNA | AB011472.1 | - | 10 | 1119 | ||
AF053977.1 | - | 10 | 1099 | |||
AF191341.1 | - | 10 | 1083 | |||
AK223250.1 | - | 10 | 1120 | |||
AK291610.1 | - | 10 | 1114 | |||
AK304635.1 | - | 9 | 996 | |||
BC017713.1 | - | 10 | 1091 | 4399056 | ||
BT009810.1 | - | 10 | 1083 |