Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108268.1 | NP_001101738.1 | 3 | 698 | ||
XM_006245996.2 | XP_006246058.1 | 4 | 609 | |||
XM_006245997.2 | XP_006246059.1 | 4 | 588 | |||
GenBank mRNA | BC167035.1 | - | 2 | 826 |