Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002819.2 | NP_001002819.2 | 16 | 1740 | ||
GenBank mRNA | BC098630.1 | - | 16 | 1740 | 7105780 |