Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108277.1 | NP_001101747.1 | 10 | 1163 | ||
XM_006246506.2 | XP_006246568.1 | 11 | 1245 | |||
XM_006246507.2 | XP_006246569.1 | 11 | 1245 | |||
XM_006246508.2 | XP_006246570.1 | 11 | 1245 | |||
XM_008767833.1 | XP_008766055.1 | 11 | 1245 | |||
XM_008767834.1 | XP_008766056.1 | 11 | 1245 | |||
XM_008767835.1 | XP_008766057.1 | 11 | 2778 |