Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001513.2 | NP_001001513.2 | 5, 6 | 674 | ||
XM_006246745.2 | XP_006246807.1 | 5, 6 | 917 | |||
GenBank mRNA | AY607588.1 | - | 5, 6 | 381 | ||
BC079107.1 | - | 5, 6 | 674 | 7125293 | ||
FQ221626.1 | - | 5, 6 | 303 |