Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnmtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108282.1 | NP_001101752.1 | 2 | 511 | ||
GenBank mRNA | FQ232324.1 | - | 2 | 509 |