Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108285.1 | NP_001101755.1 | 6, 7 | 869 | ||
XM_006246907.1 | XP_006246969.1 | 6, 7 | 1069 |