Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079893.2 | NP_001073362.1 | 2 | 455 | ||
NM_001270835.1 | NP_001257764.1 | 2 | 442 | |||
NM_001270836.1 | NP_001257765.1 | 2 | 446 | |||
GenBank mRNA | BC158555.1 | - | 2 | 424 | 7311099 | |
EF122004.1 | - | 1 | 268 | |||
EF122005.1 | - | 1 | 268 | |||
FQ213995.1 | - | 2 | 442 |
Gene Symbol: Dusp14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749277.3 | XP_003749325.1 | 1 | 481 |