Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004273.1 | NP_001004273.1 | 12 | 2166 | ||
XM_006247049.2 | XP_006247111.1 | 14 | 2377 | |||
XM_006247050.2 | XP_006247112.1 | 14 | 2370 | |||
XM_006247053.2 | XP_006247115.1 | 13 | 2249 | |||
XM_006247054.2 | XP_006247116.1 | 13 | 2242 | |||
GenBank mRNA | AY633742.1 | - | 12 | 2166 |