Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247233.2 | XP_006247295.1 | 3 | 444 | ||
XM_006247234.2 | XP_006247296.1 | 3 | 448 | |||
XM_006247235.2 | XP_006247297.1 | 3 | 443 |