Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mien1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108296.1 | NP_001101766.1 | 4 | 515 | ||
GenBank mRNA | FQ214212.1 | - | 512 |
Gene Symbol: LOC102552549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247244.2 | XP_006247306.1 | 4 | 828 |