Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam134c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135804.1 | NP_001129276.1 | 6 | 680 | ||
GenBank mRNA | BC166581.1 | - | 6 | 680 | 7380672 | |
FQ212358.1 | - | 6 | 797 |