Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037202.1 | NP_001032279.1 | 7 | 1318 | ||
XM_006247363.2 | XP_006247425.1 | 7 | 1305 | |||
XM_006247364.2 | XP_006247426.1 | 7 | 1305 | |||
XM_006247365.2 | XP_006247427.1 | 7 | 1379 | |||
GenBank mRNA | BC107561.1 | - | 7 | 1318 | 7937018 |