Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003786.3 | NP_003777.2 | 30 | 4422 | ||
XM_005257763.3 | XP_005257820.1 | 29 | 4244 | |||
XM_011525422.2 | XP_011523724.1 | 29 | 4349 | |||
XM_011525423.1 | XP_011523725.1 | 30 | 4541 | |||
XM_011525424.2 | XP_011523726.1 | 25 | 3772 | |||
XM_011525425.2 | XP_011523727.1 | 29 | 4249 | |||
XM_017025265.1 | XP_016880754.1 | 26 | 4285 | |||
GenBank mRNA | AB010887.1 | - | 30 | 4356 | ||
AB208954.1 | - | 30 | 4362 | |||
AF009670.1 | - | 30 | 4415 | |||
AF083552.1 | - | 30 | 4359 | |||
AF085690.1 | - | 30 | 4378 | |||
AF085691.1 | - | 29 | 4527 | |||
AF085692.1 | - | 29 | 4582 | |||
AF104943.1 | - | 30 | 4346 | |||
AF154001.1 | - | 5 | 1487 | |||
BC137347.1 | - | 30 | 4373 | 9021345 | ||
BC137348.1 | - | 30 | 4373 | 9021346 | ||
U66674.1 | - | 8 | 1215 | |||
U83659.1 | - | 8 | 1218 | |||
Y17151.2 | - | 30 | 4384 |