Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198547.1 | NP_001185476.1 | 8 | 1469 | ||
NM_001198549.1 | NP_001185478.1 | 4 | 684 | |||
NM_003787.4 | NP_003778.2 | 8 | 1580 | |||
XM_006722563.2 | XP_006722626.1 | 9 | 2337 | |||
XM_006722565.1 | XP_006722628.1 | 9 | 1356 | |||
XM_011526237.1 | XP_011524539.1 | 9 | 2334 | |||
XM_011526238.1 | XP_011524540.1 | 8 | 1946 | |||
XM_011526239.2 | XP_011524541.1 | 7 | 1338 | |||
XM_017026052.1 | XP_016881541.1 | 7 | 1168 | |||
XM_017026053.1 | XP_016881542.1 | 8 | 1320 | |||
XM_017026054.1 | XP_016881543.1 | 9 | 1448 | |||
XM_017026057.1 | XP_016881546.1 | 9 | 1912 | |||
XM_017026058.1 | XP_016881547.1 | 8 | 1059 | |||
XM_017026059.1 | XP_016881548.1 | 9 | 2927 | |||
GenBank mRNA | AB015339.1 | - | 3 | 277 | ||
AB017800.1 | - | 8 | 1590 | |||
AK296539.1 | - | 4 | 667 | |||
AK299850.1 | - | 8 | 1091 |