Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012143.2 | NP_001012143.2 | 2 | 753 | ||
XM_006247806.1 | XP_006247868.1 | 2 | 586 | |||
GenBank mRNA | BC079012.1 | - | 2 | 753 | 7113380 |