Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271364.1 | NP_001258293.1 | 3, 4 | 382 | ||
XM_006247861.2 | XP_006247923.1 | 3, 4 | 443 | |||
XM_006247864.2 | XP_006247926.1 | 3, 4 | 443 | |||
XM_008768500.1 | XP_008766722.1 | 3, 4 | 443 | |||
XM_008768501.1 | XP_008766723.1 | 3, 4 | 386 |