Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281297.1 | NP_001268226.1 | 4 | 397 | ||
NM_001281298.1 | NP_001268227.1 | 4 | 334 | |||
NM_001281299.1 | NP_001268228.1 | 4 | 343 | |||
NM_003792.3 | NP_003783.1 | 4 | 397 | |||
NM_153200.2 | NP_694880.1 | 4 | 397 | |||
GenBank mRNA | AB002282.1 | - | 4 | 356 | ||
AB002283.2 | - | 4 | 356 | |||
BC015500.1 | - | 4 | 347 | 3903539 | ||
BI833652.1 | - | 4 | 316 | |||
BM800379.1 | - | 4 | 359 | |||
BQ279031.1 | - | 4 | 346 | |||
BT009863.1 | - | 4 | 319 | |||
CR541914.1 | - | 4 | 319 | |||
HY029111.1 | - | 4 | 397 | |||
HY086994.1 | - | 4 | 280 | |||
HY170122.1 | - | 4 | 311 |