Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108315.1 | NP_001101785.1 | 7 | 757 | ||
XM_008768582.1 | XP_008766804.1 | 7 | 752 |