Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300928.1 | NP_001287857.1 | 2 | 537 | ||
NM_001300929.1 | NP_001287858.1 | 2 | 471 | |||
NM_003795.5 | NP_003786.1 | 2 | 537 | |||
GenBank mRNA | AF034546.1 | - | 2 | 241 | ||
AF062483.1 | - | 2 | 217 | |||
AK289476.1 | - | 2 | 328 | |||
AV704272.1 | - | 2 | 511 | |||
BC008444.1 | - | 2 | 295 | 4102735 | ||
BC014580.1 | - | 2 | 319 | 3932208 | ||
BC015179.1 | - | 2 | 301 | 3905515 | ||
BC016863.2 | - | 2 | 551 | 3846511 | ||
BG497532.1 | - | 2 | 535 | |||
BT007114.1 | - | 2 | 198 | |||
CN422502.1 | - | 2 | 217 | |||
CR456898.1 | - | 2 | 198 | |||
HY156437.1 | - | 2 | 375 |