Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012146.1 | NP_001012146.1 | 7 | 1147 | ||
XM_008768890.1 | XP_008767112.1 | 7 | 1130 | |||
GenBank mRNA | BC081905.1 | - | 7 | 1147 | 7111034 | |
FQ211720.1 | - | 7 | 1071 | |||
FQ228410.1 | - | 7 | 1049 |
Gene Symbol: LOC100910275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750241.2 | XP_003750289.1 | 7 | 1146 | ||
XM_006240675.2 | XP_006240737.1 | 7 | 1123 |
Gene Symbol: LOC103693608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768936.1 | XP_008767158.1 | 1 | 270 |