Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108336.1 | NP_001101806.1 | 7, 8 | 1054 | ||
XM_006249399.2 | XP_006249461.1 | 5, 6 | 1035 |