Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249420.2 | XP_006249482.1 | 25 | 5627 | ||
XM_006249421.2 | XP_006249483.1 | 26 | 6004 | |||
XM_006249422.2 | XP_006249484.1 | 26 | 5977 | |||
GenBank mRNA | GU566026.1 | - | 26 | 5965 |