Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suds3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769301.1 | XP_008767523.1 | 3, 4 | 410 | ||
XM_341092.7 | XP_341093.4 | 3, 4 | 409 |