Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199378.1 | NP_001186307.1 | 3 | 422 | ||
NM_001199379.1 | NP_001186308.1 | 3 | 422 | |||
NM_004193.2 | NP_004184.1 | 3 | 422 | |||
XM_005270261.1 | XP_005270318.1 | 3 | 422 | |||
XM_006718047.1 | XP_006718110.1 | 3 | 422 | |||
XM_006718048.1 | XP_006718111.1 | 3 | 422 | |||
XM_006718049.1 | XP_006718112.1 | 3 | 422 | |||
XM_006718050.1 | XP_006718113.1 | 3 | 422 | |||
XM_011540312.1 | XP_011538614.1 | 3 | 422 | |||
XM_011540313.1 | XP_011538615.1 | 3 | 422 | |||
XM_017016861.1 | XP_016872350.1 | 3 | 422 | |||
XM_017016862.1 | XP_016872351.1 | 3 | 422 | |||
XM_017016863.1 | XP_016872352.1 | 3 | 422 | |||
GenBank mRNA | AF068755.1 | - | 3 | 368 | ||
AK025330.1 | - | 3 | 388 | |||
BC032543.1 | - | 3 | 382 | |||
BC094763.1 | - | 3 | 389 | |||
BC117681.1 | - | 3 | 249 | 40070852 | ||
BC117682.1 | - | 3 | 249 | 40070853 | ||
D87435.2 | - | 3 | 352 | |||
DA176540.1 | - | 3 | 400 | |||
DA772398.1 | - | 3 | 422 |