Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199378.1 | NP_001186307.1 | 35 | 5002 | ||
NM_001199379.1 | NP_001186308.1 | 35 | 4999 | |||
NM_004193.2 | NP_004184.1 | 35 | 5011 | |||
XM_005270261.1 | XP_005270318.1 | 35 | 5014 | |||
XM_006718047.1 | XP_006718110.1 | 36 | 5113 | |||
XM_006718048.1 | XP_006718111.1 | 36 | 5110 | |||
XM_006718049.1 | XP_006718112.1 | 36 | 5101 | |||
XM_006718050.1 | XP_006718113.1 | 36 | 5077 | |||
XM_011540312.1 | XP_011538614.1 | 35 | 4972 | |||
XM_011540313.1 | XP_011538615.1 | 36 | 5113 | |||
XM_011540314.2 | XP_011538616.1 | 24 | 3491 | |||
XM_017016861.1 | XP_016872350.1 | 36 | 5089 | |||
XM_017016862.1 | XP_016872351.1 | 36 | 5065 | |||
XM_017016863.1 | XP_016872352.1 | 35 | 4975 | |||
XM_017016864.1 | XP_016872353.1 | 31 | 4313 | |||
GenBank mRNA | AF068755.1 | - | 35 | 4957 | ||
AK024916.1 | - | 15 | 2112 | |||
AK025153.1 | - | 8 | 1275 | |||
AK025330.1 | - | 35 | 4968 | |||
BC007941.2 | - | 1 | 31 | |||
BC014171.2 | - | 5 | 785 | |||
BC117681.1 | - | 35 | 4826 | 40070852 | ||
BC117682.1 | - | 35 | 4826 | 40070853 | ||
D87435.2 | - | 35 | 4941 |