Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077826.1 | NP_001071294.1 | 3 | 339 | ||
GenBank mRNA | BC158557.1 | - | 442 | 7313337 | ||
EF044301.1 | - | 3 | 339 | |||
FQ213412.1 | - | 3 | 350 |