Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108359.1 | NP_001101829.1 | 8 | 634 | ||
XM_006250907.2 | XP_006250969.1 | 6 | 533 | |||
XM_008770158.1 | XP_008768380.1 | 8 | 645 | |||
XM_008770159.1 | XP_008768381.1 | 6 | 436 |