Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003809.2 | NP_003800.1 | 6, 7 | 593 | ||
NR_037146.1 | 7, 8 | 832 | ||||
GenBank mRNA | AF030099.1 | - | 6, 7 | 514 | ||
AF055872.1 | - | 6, 7 | 554 | |||
AY358870.1 | - | 6, 7 | 554 | |||
BC019047.2 | - | 7, 8 | 832 | 4766071 | ||
BC071837.1 | - | 7, 8 | 815 | 5934943 | ||
BC104420.1 | - | 6, 7 | 529 | 40004935 |