Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142371.1 | NP_001135843.1 | 28 | 2249 | ||
GenBank mRNA | BC092603.1 | - | 17 | 1243 | ||
BC160927.1 | - | 15 | 1036 |