Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108367.1 | NP_001101837.1 | 10 | 1835 | ||
GenBank mRNA | FQ211609.1 | - | 7 | 1113 |