Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108369.1 | NP_001101839.1 | 2 | 244 | ||
GenBank mRNA | BC168773.1 | - | 2 | 240 | 6920789 | |
FQ217372.1 | - | 2 | 217 | |||
FQ217729.1 | - | 2 | 224 | |||
FQ221927.1 | - | 2 | 210 | |||
FQ224582.1 | - | 2 | 225 | |||
FQ224809.1 | - | 2 | 225 |