Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024272.1 | NP_001019443.1 | 9 | 744 | ||
GenBank mRNA | BC088433.1 | - | 9 | 744 | 7124805 | |
FQ230234.1 | - | 5 | 324 | |||
FQ232641.1 | - | 5 | 324 |