Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034937.2 | NP_001030109.2 | 7, 8 | 941 | ||
XM_006252509.2 | XP_006252571.1 | 6, 7 | 967 | |||
GenBank mRNA | BC105842.1 | - | 2, 3 | 324 | 7455960 |