Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003816.2 | NP_003807.1 | 16 | 1840 | ||
NR_027638.1 | 16 | 1840 | ||||
NR_027639.1 | 16 | 1840 | ||||
NR_027878.1 | 16 | 1840 | ||||
XM_011544682.2 | XP_011542984.1 | 16 | 2090 | |||
XM_017013942.1 | XP_016869431.1 | 16 | 1783 | |||
GenBank mRNA | AF495383.1 | - | 16 | 1795 | ||
AK293257.1 | - | 3 | 206 | |||
BC126406.1 | - | 16 | 1800 | 8992122 | ||
BC143923.1 | - | 16 | 1800 | 9052441 | ||
BC143924.1 | - | 16 | 1800 | |||
D14665.2 | - | 12 | 1415 | |||
U41766.1 | - | 16 | 1840 |