Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108397.1 | NP_001101867.1 | 7, 8 | 3310 | ||
XM_008771124.1 | XP_008769346.1 | 6, 7 | 2897 | |||
XM_008771125.1 | XP_008769347.1 | 7, 8 | 2937 | |||
XM_008771126.1 | XP_008769348.1 | 7, 8 | 3072 | |||
XM_008771127.1 | XP_008769349.1 | 6, 7 | 2894 | |||
XM_008771128.1 | XP_008769350.1 | 7, 8 | 2972 |