Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108397.1 | NP_001101867.1 | 3 | 1554 | ||
XM_008771124.1 | XP_008769346.1 | 2 | 1141 | |||
XM_008771125.1 | XP_008769347.1 | 3 | 1181 | |||
XM_008771126.1 | XP_008769348.1 | 3 | 1316 | |||
XM_008771127.1 | XP_008769349.1 | 2 | 1141 | |||
XM_008771128.1 | XP_008769350.1 | 3 | 1216 |