Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014142.1 | NP_001014164.1 | 12 | 1401 | ||
XM_006253159.2 | XP_006253221.1 | 12 | 1391 | |||
XM_008771265.1 | XP_008769487.1 | 12 | 1552 | |||
GenBank mRNA | BC083771.1 | - | 12 | 1401 | 7193395 |