Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003818.3 | NP_003809.1 | 5, 6 | 855 | ||
XM_006723660.1 | XP_006723723.1 | 5, 6 | 846 | |||
XM_011529390.1 | XP_011527692.1 | 6, 7 | 1178 | |||
GenBank mRNA | AF069532.1 | - | 4, 5 | 451 | ||
AK001414.1 | - | 5, 6 | 576 | |||
AK027695.1 | - | 5, 6 | 562 | |||
AK091045.1 | - | 6, 7 | 1178 | |||
AK095430.1 | - | 2, 3 | 779 | |||
AK303426.1 | - | 1, 2 | 129 | |||
AK304763.1 | - | 2, 3 | 319 | |||
AK315489.1 | - | 5, 6 | 836 | |||
BC025751.1 | - | 5, 6 | 576 | 5199748 | ||
Y16521.1 | - | 5, 6 | 781 |