Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003818.3 | NP_003809.1 | 13 | 1557 | ||
XM_006723660.1 | XP_006723723.1 | 13 | 1568 | |||
XM_011529390.1 | XP_011527692.1 | 14 | 1880 | |||
GenBank mRNA | AF069532.1 | - | 12 | 1153 | ||
AK001414.1 | - | 13 | 1278 | |||
AK027695.1 | - | 13 | 1264 | |||
AK095430.1 | - | 10 | 1481 | |||
AK304763.1 | - | 10 | 1021 | |||
AK315489.1 | - | 13 | 1538 | |||
BC025751.1 | - | 13 | 1278 | 5199748 | ||
Y16521.1 | - | 13 | 1483 |