Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253460.2 | XP_006253522.1 | 1 | 218 | ||
XM_006253461.2 | XP_006253523.1 | 1 | 216 | |||
XM_006253462.2 | XP_006253524.1 | 1 | 209 |