Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024796.1 | NP_001019967.1 | 587 | |||
GenBank mRNA | BC097359.1 | - | 587 | 7442223 |
Gene Symbol: LOC100363675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252535.1 | XP_006252597.1 | 4 | 581 | ||
XM_008771015.1 | XP_008769237.1 | 3 | 5693 |