Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297605.1 | NP_001284534.1 | 7 | 1435 | ||
NM_003820.3 | NP_003811.2 | 8 | 1578 | |||
XM_006711019.2 | XP_006711082.1 | 8 | 1250 | |||
XM_011542383.2 | XP_011540685.1 | 5 | 1621 | |||
XM_017002718.1 | XP_016858207.1 | 5 | 1701 | |||
XM_017002720.1 | XP_016858209.1 | 8 | 2296 | |||
XM_017002721.1 | XP_016858210.1 | 9 | 1706 | |||
GenBank mRNA | AB208808.1 | - | 5 | 4205 | ||
AF153978.1 | - | 8 | 1523 | |||
AK124010.1 | - | 6 | 2477 | |||
BC002794.2 | - | 8 | 1544 | 3614650 | ||
BC018882.2 | - | 8 | 1544 | |||
BC029848.1 | - | 6 | 1515 | 5178561 | ||
U70321.1 | - | 8 | 1571 | |||
U81232.1 | - | 8 | 1542 |