Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024273.1 | NP_001019444.1 | 6 | 781 | ||
GenBank mRNA | BC095843.1 | - | 6 | 781 | 7389569 |