Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108417.1 | NP_001101887.1 | 4, 5 | 505 | ||
XM_006254072.2 | XP_006254134.1 | 4, 5 | 500 | |||
XM_006254073.2 | XP_006254135.1 | 4, 5 | 505 | |||
XM_006254074.2 | XP_006254136.1 | 4, 5 | 421 |