Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108418.1 | NP_001101888.1 | 906 | |||
XM_006254175.2 | XP_006254237.1 | 1057 | ||||
XM_006254176.2 | XP_006254238.1 | 1072 | ||||
XM_008771752.1 | XP_008769974.1 | 1340 |
Gene Symbol: NEWGENE_1564769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254170.2 | XP_006254232.1 | 7 | 1057 | ||
XM_006254171.2 | XP_006254233.1 | 7 | 1072 | |||
XM_008771835.1 | XP_008770057.1 | 6 | 1340 |