Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003825.3 | NP_003816.2 | 8 | 1012 | ||
NM_130798.2 | NP_570710.1 | 7 | 853 | |||
XM_006720725.2 | XP_006720788.1 | 9 | 1091 | |||
GenBank mRNA | AJ011915.1 | - | 8 | 634 | ||
AK290152.1 | - | 8 | 1058 | |||
AK313161.1 | - | 8 | 656 | |||
AK315279.1 | - | 8 | 911 | |||
BC000148.2 | - | 8 | 644 | 2900640 | ||
BC003686.1 | - | 8 | 663 | 3461227 | ||
BC022890.1 | - | 8 | 668 | 4658930 | ||
BT006916.1 | - | 7 | 590 | |||
CR457212.1 | - | 7 | 590 | |||
DA650133.1 | - | 7 | 481 | |||
U55936.1 | - | 8 | 682 | |||
Y09567.1 | - | 7 | 590 | |||
Y09568.1 | - | 6 | 431 |